Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAGGAAGAAAATCAATGTGAGA[C/G]ACTCTTCACCATCACATGGGCAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616706 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARP11 - poly(ADP-ribose) polymerase family member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286521.1 | 2352 | UTR 3 | NP_001273450.1 | |||
NM_001286522.1 | 2352 | UTR 3 | NP_001273451.1 | |||
NM_020367.5 | 2352 | UTR 3 | NP_065100.2 | |||
XM_005253711.4 | 2352 | Intron | XP_005253768.1 | |||
XM_011520970.1 | 2352 | Intron | XP_011519272.1 | |||
XM_011520971.2 | 2352 | Intron | XP_011519273.1 | |||
XM_011520973.2 | 2352 | Intron | XP_011519275.1 | |||
XM_017019670.1 | 2352 | Intron | XP_016875159.1 | |||
XM_017019671.1 | 2352 | Intron | XP_016875160.1 |