Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAGGAAACTTGGGGGTAATGGA[C/T]GTTTTTATTATCGTGATTGTAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601430 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UPF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UPF1 - UPF1, RNA helicase and ATPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297549.1 | Intron | NP_001284478.1 | ||||
NM_002911.3 | Intron | NP_002902.2 | ||||
XM_017027105.1 | Intron | XP_016882594.1 | ||||
XM_017027106.1 | Intron | XP_016882595.1 |
Set Membership: |
HapMap |