Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCTGTACAGTTTTGTTTTACTT[C/T]TGTTTCCGGCTACTTTTCACTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611567 MIM: 614289 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MACROD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MACROD2 - MACRO domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033087.1 | Intron | NP_001028259.1 | ||||
NM_080676.5 | Intron | NP_542407.2 | ||||
XM_017027675.1 | Intron | XP_016883164.1 | ||||
XM_017027676.1 | Intron | XP_016883165.1 | ||||
XM_017027677.1 | Intron | XP_016883166.1 |
SEL1L2 - SEL1L2 ERAD E3 ligase adaptor subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |