Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609803 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKAR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OSGEPL1 - O-sialoglycoprotein endopeptidase-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022353.2 | Intron | NP_071748.2 | ||||
XM_005246766.3 | Intron | XP_005246823.1 | ||||
XM_006712685.1 | Intron | XP_006712748.1 | ||||
XM_006712686.1 | Intron | XP_006712749.1 | ||||
XM_011511631.1 | Intron | XP_011509933.1 | ||||
XM_017004676.1 | Intron | XP_016860165.1 | ||||
XM_017004677.1 | Intron | XP_016860166.1 | ||||
XM_017004678.1 | Intron | XP_016860167.1 | ||||
XM_017004679.1 | Intron | XP_016860168.1 | ||||
XM_017004680.1 | Intron | XP_016860169.1 | ||||
XM_017004681.1 | Intron | XP_016860170.1 | ||||
XM_017004682.1 | Intron | XP_016860171.1 | ||||
XM_017004683.1 | Intron | XP_016860172.1 | ||||
XM_017004684.1 | Intron | XP_016860173.1 | ||||
XM_017004685.1 | Intron | XP_016860174.1 |