Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTTTCTCCTTCATCGCTGGTGT[C/G]CCAACAAAAAGAGTAAGTCTCACGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604938 MIM: 610095 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIR2DL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIR2DL3 - killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIR3DL3 - killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153443.4 | 1092 | Missense Mutation | CCC,GCC | P,A 348 | NP_703144.3 |
Set Membership: |
HapMap |