Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCGTCCATGGCACCCTGCCAGG[C/T]GCGTGCCTGGTGCTCCTCTAGCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608066 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAFB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SAFB2 - scaffold attachment factor B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014649.2 | 2143 | Missense Mutation | NP_055464.1 | |||
XM_011528449.2 | 2143 | Missense Mutation | XP_011526751.1 | |||
XM_011528450.2 | 2143 | Missense Mutation | XP_011526752.1 | |||
XM_017027520.1 | 2143 | Missense Mutation | XP_016883009.1 | |||
XM_017027521.1 | 2143 | Missense Mutation | XP_016883010.1 | |||
XM_017027522.1 | 2143 | Missense Mutation | XP_016883011.1 |