Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAATATTTTCTCTAGCAACTAA[C/T]GAGATGTGGCGATCAGGGGATAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600439 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SSBP1 - single stranded DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256510.1 | 447 | Silent Mutation | AAC,AAT | N,N 62 | NP_001243439.1 | |
NM_001256511.1 | 447 | Silent Mutation | AAC,AAT | N,N 62 | NP_001243440.1 | |
NM_001256512.1 | 447 | Silent Mutation | AAC,AAT | N,N 62 | NP_001243441.1 | |
NM_001256513.1 | 447 | Intron | NP_001243442.1 | |||
NM_003143.2 | 447 | Intron | NP_003134.1 | |||
XM_005250048.4 | 447 | Silent Mutation | AAC,AAT | N,N 62 | XP_005250105.1 | |
XM_005250049.4 | 447 | Silent Mutation | AAC,AAT | N,N 62 | XP_005250106.1 | |
XM_005250050.4 | 447 | Silent Mutation | AAC,AAT | N,N 62 | XP_005250107.1 | |
XM_005250051.4 | 447 | Silent Mutation | AAC,AAT | N,N 62 | XP_005250108.1 |
WEE2-AS1 - WEE2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |