Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCTTCTTCTGCCTGCGACAGAC[C/T]ACCAGCCCCCGGAACTGGTGCATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608230 | ||||||||||||||||||||
Literature Links: |
CACNA1I PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNA1I - calcium voltage-gated channel subunit alpha1 I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003406.1 | 195 | Silent Mutation | ACC,ACT | T,T 65 | NP_001003406.1 | |
NM_021096.3 | 195 | Silent Mutation | ACC,ACT | T,T 65 | NP_066919.2 | |
XM_017029035.1 | 195 | Intron | XP_016884524.1 | |||
XM_017029036.1 | 195 | Intron | XP_016884525.1 | |||
XM_017029037.1 | 195 | Intron | XP_016884526.1 |