Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTCACTTCAATGCCCCCTCACAC[A/G]TGCGCAGGAAGATCATGTCATCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERGIC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERGIC1 - endoplasmic reticulum-golgi intermediate compartment 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100268168 - uncharacterized LOC100268168 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL26L1 - ribosomal protein L26 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317980.1 | 169 | Missense Mutation | ATG,GTG | M,V 25 | NP_001304909.1 | |
NM_001317981.1 | 169 | Missense Mutation | ATG,GTG | M,V 25 | NP_001304910.1 | |
NM_001317982.1 | 169 | Missense Mutation | ATG,GTG | M,V 25 | NP_001304911.1 | |
NM_016093.3 | 169 | Intron | NP_057177.1 | |||
XM_005265921.3 | 169 | Missense Mutation | ATG,GTG | M,V 54 | XP_005265978.1 | |
XM_011534565.2 | 169 | Missense Mutation | ATG,GTG | M,V 25 | XP_011532867.1 | |
XM_017009519.1 | 169 | Missense Mutation | ATG,GTG | M,V 25 | XP_016865008.1 | |
XM_017009520.1 | 169 | Intron | XP_016865009.1 |