Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATGTCTTTGAAAACAATGAAGG[G/A]GACGGTAATAACTTAGAGTGAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608124 | ||||||||||||||||||||
Literature Links: |
XYLT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
XYLT1 - xylosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022166.3 | 8775 | UTR 3 | NP_071449.1 | |||
XM_017023539.1 | 8775 | Intron | XP_016879028.1 | |||
XM_017023540.1 | 8775 | Intron | XP_016879029.1 |