Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCAGGAGAGCAGCCGTCTCCTT[C/G]TCGGGGGCAGGACCAGGTGCCGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMC7 - armadillo repeat containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304271.1 | 495 | Intron | NP_001291200.1 | |||
NM_001304272.1 | 495 | Intron | NP_001291201.1 | |||
NM_001304273.1 | 495 | Intron | NP_001291202.1 | |||
NM_024585.3 | 495 | Intron | NP_078861.1 |
HN1 - hematological and neurological expressed 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NT5C - 5', 3'-nucleotidase, cytosolic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252377.1 | 495 | Missense Mutation | ACA,AGA | T,R 136 | NP_001239306.1 | |
NM_014595.2 | 495 | Missense Mutation | ACA,AGA | T,R 176 | NP_055410.1 | |
XM_011524700.2 | 495 | Intron | XP_011523002.2 |