Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGGGAGATCAGTCCTTAATTA[C/T]GGATACAGTGAACTCAAATGGAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611688 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KHDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KHDC1 - KH homology domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251874.1 | 928 | Silent Mutation | CCA,CCG | P,P 237 | NP_001238803.1 | |
NM_030568.4 | 928 | Silent Mutation | CCA,CCG | P,P 164 | NP_085045.3 |