Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTAAGGTTGCTCCCCAGTATGAA[A/T]TCTTAAATGTCTTACAAGGCATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF836 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF836 - zinc finger protein 836 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102657.1 | 2048 | Missense Mutation | ATT,TTT | I,F 547 | NP_001096127.1 | |
XM_011526558.2 | 2048 | Missense Mutation | ATT,TTT | I,F 547 | XP_011524860.1 | |
XM_011526559.2 | 2048 | Missense Mutation | ATT,TTT | I,F 547 | XP_011524861.1 | |
XM_017026389.1 | 2048 | Intron | XP_016881878.1 |