Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCGCTGTGCCTGCTGCTGAGCA[C/G]CGCCGCGGGTGAGTCGGGGGGCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDLRAD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LDLRAD3 - low density lipoprotein receptor class A domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304263.1 | 156 | Missense Mutation | ACC,AGC | T,S 13 | NP_001291192.1 | |
NM_001304264.1 | 156 | UTR 5 | NP_001291193.1 | |||
NM_174902.3 | 156 | Missense Mutation | ACC,AGC | T,S 13 | NP_777562.1 |