Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTTAGGAAGGTGCTTCATGGCA[C/T]TGCTACGGAGACAGGCGATGTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609506 MIM: 613336 MIM: 604466 MIM: 615258 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP27B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP27B1 - cytochrome P450 family 27 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MARCH9 - membrane associated ring-CH-type finger 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL1 - methyltransferase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005371.5 | 585 | Missense Mutation | AAT,AGT | N,S 140 | NP_005362.3 | |
NM_023033.3 | 585 | Intron | NP_075422.3 | |||
XM_005268873.2 | 585 | Missense Mutation | AAT,AGT | N,S 97 | XP_005268930.1 | |
XM_017019305.1 | 585 | Missense Mutation | AAT,AGT | N,S 165 | XP_016874794.1 |
METTL21B - methyltransferase like 21B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |