Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAACTGGGGGTGGTGGCGGTGGG[A/G]AAGACAGGGAAGATTGTAGAGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605220 MIM: 607042 MIM: 608273 | ||||||||||||||||||||
Literature Links: |
APOBR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOBR - apolipoprotein B receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018690.3 | 192 | Silent Mutation | GGA,GGG | G,G 53 | NP_061160.3 |
CLN3 - CLN3, battenin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL27 - interleukin 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |