Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGTTGGGGGATGGTGTCTATGAC[A/G]CCTTCATGATGATAGATGAAACCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603497 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYRK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYRK3 - dual specificity tyrosine phosphorylation regulated kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004023.1 | 362 | Missense Mutation | ACC,GCC | T,A 14 | NP_001004023.1 | |
NM_003582.2 | 362 | Missense Mutation | ACC,GCC | T,A 34 | NP_003573.2 | |
XM_005273315.4 | 362 | Missense Mutation | ACC,GCC | T,A 14 | XP_005273372.1 | |
XM_011510061.2 | 362 | UTR 5 | XP_011508363.1 |