Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTTTCCACACTCCTTACACTGA[A/G]AGGGTTTCTCTCCTGTATGGATTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194527 | ||||||||||||||||||||
Literature Links: |
ZNF23 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF23 - zinc finger protein 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304492.1 | 2502 | Intron | NP_001291421.1 | |||
NM_001304493.1 | 2502 | Missense Mutation | TCT,TTT | S,F 502 | NP_001291422.1 | |
NM_001304494.1 | 2502 | Intron | NP_001291423.1 | |||
NM_145911.2 | 2502 | Missense Mutation | TCT,TTT | S,F 560 | NP_666016.1 |