Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGCTGGGGCGCACCGAGACCA[C/T]GTCTGCGATGCAGCGGTTGAGGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607059 MIM: 604546 MIM: 611952 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC39A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC39A4 - solute carrier family 39 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TONSL - tonsoku-like, DNA repair protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS28 - VPS28, ESCRT-I subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016208.3 | 503 | Missense Mutation | ATG,GTG | M,V 132 | NP_057292.1 | |
NM_183057.2 | 503 | Missense Mutation | ATG,GTG | M,V 132 | NP_898880.1 | |
XM_005272323.3 | 503 | Missense Mutation | ATG,GTG | M,V 115 | XP_005272380.1 | |
XM_005272324.3 | 503 | Missense Mutation | ATG,GTG | M,V 115 | XP_005272381.1 | |
XM_005272325.3 | 503 | Missense Mutation | ATG,GTG | M,V 94 | XP_005272382.1 | |
XM_011517099.1 | 503 | Missense Mutation | ATG,GTG | M,V 94 | XP_011515401.1 | |
XM_011517100.1 | 503 | Missense Mutation | ATG,GTG | M,V 94 | XP_011515402.1 | |
XM_017013529.1 | 503 | Missense Mutation | ATG,GTG | M,V 94 | XP_016869018.1 |