Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTAGTCAGCGGAGGACCAGAGTG[A/G]GTTAAAATTGGTTTCAGATACCTGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608396 | |||||||||||||||||||||||
Literature Links: |
SLC9A9 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
SLC9A9 - solute carrier family 9 member A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173653.3 | 1835 | Silent Mutation | NP_775924.1 | |||
XM_011512703.2 | 1835 | Silent Mutation | XP_011511005.1 | |||
XM_011512704.2 | 1835 | Intron | XP_011511006.1 | |||
XM_017006202.1 | 1835 | Intron | XP_016861691.1 | |||
XM_017006203.1 | 1835 | Silent Mutation | XP_016861692.1 |