Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGACCTCTCGCCTGCTTCTTGTG[A/G]TTCTGAGCACCTCTAGATAACTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COPS9 - COP9 signalosome subunit 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163424.1 | 1033 | Intron | NP_001156896.1 | |||
NM_138336.1 | 1033 | Missense Mutation | ACC,ATC | T,I 203 | NP_612209.1 | |
XM_017003411.1 | 1033 | Missense Mutation | ACC,ATC | T,I 193 | XP_016858900.1 |