Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTTCAGCTTCATTAAGACCATG[A/G]TGATCCTCTTCAATTTGCTCATCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606076 MIM: 606822 MIM: 613170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3R3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIK3R3 - phosphoinositide-3-kinase regulatory subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POMGNT1 - protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPAN1 - tetraspanin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005727.3 | 479 | Missense Mutation | ATG,GTG | M,V 11 | NP_005718.2 | |
XM_011540460.2 | 479 | Missense Mutation | ATG,GTG | M,V 11 | XP_011538762.1 |