Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATGATCACTTTGGTGAAGAAGG[C/T]GTCAGATACTATTTTATTCTGAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613946 MIM: 602878 MIM: 606474 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC5G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC5G - DnaJ heat shock protein family (Hsp40) member C5 gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303127.1 | 692 | Silent Mutation | GGC,GGT | G,G 112 | NP_001290056.1 | |
NM_001303128.1 | 692 | Intron | NP_001290057.1 | |||
NM_173650.2 | 692 | Silent Mutation | GGC,GGT | G,G 112 | NP_775921.1 | |
XM_006711995.3 | 692 | Silent Mutation | GGC,GGT | G,G 112 | XP_006712058.1 | |
XM_017003872.1 | 692 | Silent Mutation | GGC,GGT | G,G 112 | XP_016859361.1 | |
XM_017003873.1 | 692 | Silent Mutation | GGC,GGT | G,G 112 | XP_016859362.1 | |
XM_017003874.1 | 692 | Silent Mutation | GGC,GGT | G,G 112 | XP_016859363.1 |
SLC30A3 - solute carrier family 30 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM54 - tripartite motif containing 54 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |