Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTGGCTCGTGCCTGGAGGACCT[A/G]GCGCGCAAGGACTACCACTACCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZCCHC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZCCHC2 - zinc finger CCHC-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017742.5 | 188 | Silent Mutation | CTA,CTG | L,L 137 | NP_060212.4 | |
XM_006722493.3 | 188 | Silent Mutation | CTA,CTG | L,L 57 | XP_006722556.1 | |
XM_011526044.1 | 188 | Intron | XP_011524346.1 | |||
XM_017025802.1 | 188 | Silent Mutation | CTA,CTG | L,L 57 | XP_016881291.1 |