Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTCCATGACAACTTTCACTGAG[C/T]CTGAAGTAGTATTCCTGCAATCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGFG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGFG2 - ArfGAP with FG repeats 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006076.4 | 402 | Missense Mutation | CCT,TCT | P,S 94 | NP_006067.3 | |
XM_005250306.2 | 402 | Missense Mutation | CCT,TCT | P,S 94 | XP_005250363.1 | |
XM_017012112.1 | 402 | Missense Mutation | CCT,TCT | P,S 7 | XP_016867601.1 | |
XM_017012113.1 | 402 | Missense Mutation | CCT,TCT | P,S 94 | XP_016867602.1 | |
XM_017012114.1 | 402 | UTR 5 | XP_016867603.1 |