Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGGCAGCCTGGGCCGGTCCGAC[C/G]GTGTGGCCAAGATGAGCCCCAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614988 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPS8L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPS8L2 - EPS8 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022772.3 | 314 | Missense Mutation | CGT,GGT | R,G 23 | NP_073609.2 | |
XM_017018130.1 | 314 | Missense Mutation | CGT,GGT | R,G 23 | XP_016873619.1 | |
XM_017018131.1 | 314 | Missense Mutation | CGT,GGT | R,G 23 | XP_016873620.1 | |
XM_017018132.1 | 314 | Missense Mutation | CGT,GGT | R,G 23 | XP_016873621.1 |
TMEM80 - transmembrane protein 80 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |