Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAACAGAAATGAAAGAAGAGGAG[A/G]AACCTAATCCAAATTATAAAGAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603146 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMD9 - proteasome 26S subunit, non-ATPase 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR66 - WD repeat domain 66 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178003.1 | 245 | Missense Mutation | AAA,GAA | K,E 25 | NP_001171474.1 | |
NM_144668.5 | 245 | Missense Mutation | AAA,GAA | K,E 25 | NP_653269.3 |