Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGATGGACCTGCCGGCACTGCTG[C/T]CAGAAGTGCTACGAGTCCAGCTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYT17 - synaptotagmin 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308157.1 | 509 | Silent Mutation | TGC,TGT | C,C 27 | NP_001295086.1 | |
NM_016524.3 | 509 | Silent Mutation | TGC,TGT | C,C 31 | NP_057608.2 | |
XM_005255362.3 | 509 | UTR 5 | XP_005255419.1 | |||
XM_011545865.1 | 509 | Silent Mutation | TGC,TGT | C,C 47 | XP_011544167.1 | |
XM_011545866.2 | 509 | Silent Mutation | TGC,TGT | C,C 38 | XP_011544168.1 | |
XM_011545868.1 | 509 | Silent Mutation | TGC,TGT | C,C 25 | XP_011544170.1 | |
XM_011545869.2 | 509 | UTR 5 | XP_011544171.1 | |||
XM_011545870.2 | 509 | UTR 5 | XP_011544172.1 | |||
XM_011545871.2 | 509 | UTR 5 | XP_011544173.1 | |||
XM_011545872.2 | 509 | UTR 5 | XP_011544174.1 | |||
XM_017023280.1 | 509 | Silent Mutation | TGC,TGT | C,C 31 | XP_016878769.1 | |
XM_017023281.1 | 509 | UTR 5 | XP_016878770.1 |