Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCGCGCCGAGCGGGAGCGGCAG[C/G]CGCACCGGCCCATAGAGCGCGCCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600126 | ||||||||||||||||||||
Literature Links: |
PDE4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDE4A - phosphodiesterase 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001111307.1 | 202 | Missense Mutation | CCG,GCG | P,A 61 | NP_001104777.1 | |
NM_001111308.1 | 202 | Intron | NP_001104778.1 | |||
NM_001111309.1 | 202 | Intron | NP_001104779.1 | |||
NM_001243121.1 | 202 | Intron | NP_001230050.1 | |||
NM_006202.2 | 202 | Intron | NP_006193.1 | |||
XM_005259933.2 | 202 | Intron | XP_005259990.1 | |||
XM_011528054.1 | 202 | Intron | XP_011526356.1 | |||
XM_011528055.2 | 202 | Missense Mutation | CCG,GCG | P,A 61 | XP_011526357.1 | |
XM_017026865.1 | 202 | Missense Mutation | CCG,GCG | P,A 61 | XP_016882354.1 |