Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCACAGTTAACTCAATATTTCC[C/T]GGAACATTCTGATTCTGCATATACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300304 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CUL4B - cullin 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079872.1 | 2286 | Silent Mutation | CCA,CCG | P,P 654 | NP_001073341.1 | |
NM_003588.3 | 2286 | Silent Mutation | CCA,CCG | P,P 672 | NP_003579.3 | |
XM_005262481.1 | 2286 | Silent Mutation | CCA,CCG | P,P 674 | XP_005262538.1 | |
XM_006724784.1 | 2286 | Silent Mutation | CCA,CCG | P,P 661 | XP_006724847.1 | |
XM_006724785.2 | 2286 | Silent Mutation | CCA,CCG | P,P 659 | XP_006724848.1 | |
XM_011531399.2 | 2286 | Silent Mutation | CCA,CCG | P,P 478 | XP_011529701.1 | |
XM_011531400.2 | 2286 | Silent Mutation | CCA,CCG | P,P 476 | XP_011529702.1 | |
XM_011531401.1 | 2286 | Silent Mutation | CCA,CCG | P,P 460 | XP_011529703.1 |