Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTCTGGACGATCTGCTTGGCAA[C/T]CTTATAGAACTGCTCCCGGTCATCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603124 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2G2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE2G2 - ubiquitin conjugating enzyme E2 G2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202489.1 | 466 | Missense Mutation | ATT,GTT | I,V 84 | NP_001189418.1 | |
NM_003343.5 | 466 | Missense Mutation | ATT,GTT | I,V 154 | NP_003334.2 | |
NM_182688.2 | 466 | Missense Mutation | ATT,GTT | I,V 126 | NP_872630.1 | |
XM_017028469.1 | 466 | Missense Mutation | ATT,GTT | I,V 126 | XP_016883958.1 |