Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCAGGTTTGGAATCCAGAGTCTT[C/T]AGATTCTTCAGTTTTTTCACTTTAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616519 | |||||||||||||||||||||||
Literature Links: |
DENND6A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DENND6A - DENN domain containing 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152678.2 | 1247 | Silent Mutation | CTA,CTG | L,L 392 | NP_689891.1 | |
XM_006713019.3 | 1247 | Silent Mutation | CTA,CTG | L,L 391 | XP_006713082.1 | |
XM_006713020.2 | 1247 | Silent Mutation | CTA,CTG | L,L 326 | XP_006713083.1 | |
XM_017005864.1 | 1247 | Silent Mutation | CTA,CTG | L,L 325 | XP_016861353.1 |
PDE12 - phosphodiesterase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322176.1 | 1247 | Intron | NP_001309105.1 | |||
NM_001322177.1 | 1247 | Intron | NP_001309106.1 | |||
NM_177966.6 | 1247 | Intron | NP_808881.3 |