Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGACCAGCGCACAGCAAACAGA[C/T]ACTTGAGGGAGAGCTGCAGCGAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182810 MIM: 613363 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPTAN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPTAN1 - spectrin alpha, non-erythrocytic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130438.2 | 1434 | Intron | NP_001123910.1 | |||
NM_001195532.1 | 1434 | Intron | NP_001182461.1 | |||
NM_003127.3 | 1434 | Intron | NP_003118.2 | |||
XM_006717245.1 | 1434 | Intron | XP_006717308.1 | |||
XM_006717246.1 | 1434 | Intron | XP_006717309.1 | |||
XM_006717247.2 | 1434 | Intron | XP_006717310.1 | |||
XM_006717248.2 | 1434 | Intron | XP_006717311.1 | |||
XM_006717249.1 | 1434 | Intron | XP_006717312.1 | |||
XM_006717250.1 | 1434 | Intron | XP_006717313.1 | |||
XM_006717251.2 | 1434 | Intron | XP_006717314.1 | |||
XM_006717252.2 | 1434 | Intron | XP_006717315.1 | |||
XM_006717253.1 | 1434 | Intron | XP_006717316.1 | |||
XM_006717254.2 | 1434 | Intron | XP_006717317.1 | |||
XM_017015059.1 | 1434 | Intron | XP_016870548.1 | |||
XM_017015060.1 | 1434 | Intron | XP_016870549.1 | |||
XM_017015061.1 | 1434 | Intron | XP_016870550.1 |
WDR34 - WD repeat domain 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052844.3 | 1434 | Missense Mutation | TAT,TGT | Y,C 437 | NP_443076.2 | |
XM_011519179.2 | 1434 | Missense Mutation | TAT,TGT | Y,C 409 | XP_011517481.1 |