Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTTCTCTTCTCCCTCTCCCAAC[C/T]TGTGTCGGGTCCTTCTTCCTGGGTA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
|||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||
Literature Links: |
HCG4 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EUR
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
EAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
SAS - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
AFR - Not Available | |||||
AMR
|
HCG4 - HLA complex group 4 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC554223 - histocompatibility antigen-related | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |