Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCAGGACAGCACCACGGTGTGAC[A/G]AAATGCAACATCACGTGCAGCAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601880 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CX3CL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CX3CL1 - C-X3-C motif chemokine ligand 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304392.1 | 198 | Intron | NP_001291321.1 | |||
NM_002996.4 | 198 | Silent Mutation | ACA,ACG | T,T 30 | NP_002987.1 |