Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGCTTTCAAATTAAGGCTGGAG[C/T]GACTGGGATAGAAAAGAGTGAGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606168 MIM: 605411 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX20 - DEAD-box helicase 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCND3 - potassium voltage-gated channel subfamily D member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004980.4 | 2239 | Missense Mutation | CAC,CGC | H,R 590 | NP_004971.2 | |
NM_172198.2 | 2239 | Missense Mutation | CAC,CGC | H,R 571 | NP_751948.1 | |
XM_005270851.4 | 2239 | Missense Mutation | CAC,CGC | H,R 590 | XP_005270908.1 | |
XM_006710629.3 | 2239 | Missense Mutation | CAC,CGC | H,R 590 | XP_006710692.1 | |
XM_006710630.3 | 2239 | Missense Mutation | CAC,CGC | H,R 571 | XP_006710693.1 | |
XM_006710631.3 | 2239 | Silent Mutation | TCA,TCG | S,S 507 | XP_006710694.1 | |
XM_006710632.3 | 2239 | Intron | XP_006710695.1 | |||
XM_011541425.2 | 2239 | Intron | XP_011539727.1 | |||
XM_011541426.2 | 2239 | Intron | XP_011539728.1 | |||
XM_011541427.2 | 2239 | Intron | XP_011539729.1 | |||
XM_011541428.2 | 2239 | Intron | XP_011539730.1 | |||
XM_017001244.1 | 2239 | Missense Mutation | CAC,CGC | H,R 590 | XP_016856733.1 | |
XM_017001245.1 | 2239 | Intron | XP_016856734.1 |