Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACATTCATATGGCTTCTCCCCCG[G/T]GTGAGTTCTTTCATGTATTTGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928464 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928464 - uncharacterized LOC101928464 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF433 - zinc finger protein 433 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080411.2 | 1153 | Missense Mutation | ACG,CCG | T,P 305 | NP_001073880.1 | |
NM_001308346.1 | 1153 | Missense Mutation | ACG,CCG | T,P 304 | NP_001295275.1 | |
NM_001308348.1 | 1153 | Missense Mutation | ACG,CCG | T,P 302 | NP_001295277.1 | |
NM_001308351.1 | 1153 | Missense Mutation | ACG,CCG | T,P 270 | NP_001295280.1 | |
NM_001308355.1 | 1153 | Missense Mutation | ACG,CCG | T,P 270 | NP_001295284.1 | |
NM_001308357.1 | 1153 | Missense Mutation | ACG,CCG | T,P 270 | NP_001295286.1 | |
XM_006722661.3 | 1153 | Missense Mutation | ACG,CCG | T,P 270 | XP_006722724.1 | |
XM_011527725.2 | 1153 | Missense Mutation | ACG,CCG | T,P 316 | XP_011526027.1 | |
XM_011527728.2 | 1153 | Missense Mutation | ACG,CCG | T,P 304 | XP_011526030.1 | |
XM_011527729.2 | 1153 | Missense Mutation | ACG,CCG | T,P 304 | XP_011526031.1 | |
XM_017026413.1 | 1153 | Intron | XP_016881902.1 |