Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGGCCCAGAAGGAGCCCGAGGAC[C/T]CCGAGGGTCAGTCCCAATGTCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604014 MIM: 600923 MIM: 604729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B4GALT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B4GALT3 - beta-1,4-galactosyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199873.1 | 1279 | Missense Mutation | AGT,GGT | S,G 348 | NP_001186802.1 | |
NM_001199874.1 | 1279 | Missense Mutation | AGT,GGT | S,G 348 | NP_001186803.1 | |
NM_003779.3 | 1279 | Missense Mutation | AGT,GGT | S,G 348 | NP_003770.1 | |
XM_005245566.1 | 1279 | Missense Mutation | AGT,GGT | S,G 348 | XP_005245623.1 | |
XM_011510093.1 | 1279 | UTR 3 | XP_011508395.1 | |||
XM_017002714.1 | 1279 | UTR 3 | XP_016858203.1 | |||
XM_017002715.1 | 1279 | Intron | XP_016858204.1 |
USP21 - ubiquitin specific peptidase 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |