Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCCAATGCTCTTGGTAGAAAGA[C/T]GCAGGATGCGACAGTCTGGAAATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR31 - WD repeat domain 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006615.2 | 1069 | Missense Mutation | ATC,GTC | I,V 160 | NP_001006616.1 | |
NM_001012361.3 | 1069 | Missense Mutation | ATC,GTC | I,V 285 | NP_001012361.1 | |
NM_145241.4 | 1069 | Missense Mutation | ATC,GTC | I,V 284 | NP_660284.1 | |
XM_011518194.2 | 1069 | Missense Mutation | ATC,GTC | I,V 260 | XP_011516496.1 |