Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCGTCTCGCAGGCAAACAGAAA[C/G]TTGCGGTCGGGCGTGACGATGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608114 MIM: 612056 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAP1 - ArfGAP with dual PH domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284308.1 | 1032 | Silent Mutation | AAC,AAG | N,K 344 | NP_001271237.1 | |
NM_001284309.1 | 1032 | Silent Mutation | AAC,AAG | N,K 261 | NP_001271238.1 | |
NM_001284310.1 | 1032 | Silent Mutation | AAC,AAG | N,K 261 | NP_001271239.1 | |
NM_001284311.1 | 1032 | Silent Mutation | AAC,AAG | N,K 238 | NP_001271240.1 | |
NM_006869.3 | 1032 | Silent Mutation | AAC,AAG | N,K 333 | NP_006860.1 |
GET4 - golgi to ER traffic protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |