Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACCAGAGACTATTTCAAGCCCTG[A/G]ATATCATATCCTGAGGGCCACAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609666 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQCD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IQCD - IQ motif containing D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPCN1 - two pore segment channel 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143819.2 | 299 | Missense Mutation | GAA,GGA | E,G 61 | NP_001137291.1 | |
NM_001301214.1 | 299 | Intron | NP_001288143.1 | |||
NM_017901.5 | 299 | UTR 5 | NP_060371.2 | |||
XM_005253906.4 | 299 | Intron | XP_005253963.1 | |||
XM_011538490.2 | 299 | Missense Mutation | GAA,GGA | E,G 61 | XP_011536792.1 | |
XM_011538492.2 | 299 | UTR 5 | XP_011536794.1 | |||
XM_011538493.2 | 299 | Intron | XP_011536795.1 | |||
XM_017019480.1 | 299 | Missense Mutation | GAA,GGA | E,G 61 | XP_016874969.1 | |
XM_017019481.1 | 299 | UTR 5 | XP_016874970.1 | |||
XM_017019482.1 | 299 | Missense Mutation | GAA,GGA | E,G 61 | XP_016874971.1 |