Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGGGGCCGCAGGGCCCCAAAGGC[A/G]CGGAGAAGCGGTACCTGTGCCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP92 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFP92 - ZFP92 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136273.1 | 703 | Missense Mutation | ACG,GCG | T,A 148 | NP_001129745.1 | |
XM_005274652.3 | 703 | Missense Mutation | ACG,GCG | T,A 148 | XP_005274709.1 | |
XM_011531115.2 | 703 | Missense Mutation | ACG,GCG | T,A 148 | XP_011529417.1 | |
XM_017029281.1 | 703 | Missense Mutation | ACG,GCG | T,A 244 | XP_016884770.1 |