Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGAGGAGGACCCGATCCTCAGC[C/T]GCACTCCGAGTCCAGTGCATCCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300227 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCML1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCML1 - sex comb on midleg-like 1 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037535.2 | 3696 | Missense Mutation | CGC,TGC | R,C 56 | NP_001032624.1 | |
NM_001037536.2 | 3696 | Missense Mutation | CGC,TGC | R,C 56 | NP_001032625.1 | |
NM_001037540.2 | 3696 | Missense Mutation | CGC,TGC | R,C 177 | NP_001032629.1 | |
NM_006746.5 | 3696 | Missense Mutation | CGC,TGC | R,C 150 | NP_006737.2 | |
XM_005274578.2 | 3696 | Missense Mutation | CGC,TGC | R,C 178 | XP_005274635.1 | |
XM_005274579.2 | 3696 | Missense Mutation | CGC,TGC | R,C 149 | XP_005274636.1 | |
XM_006724508.3 | 3696 | Missense Mutation | CGC,TGC | R,C 177 | XP_006724571.1 | |
XM_006724509.3 | 3696 | Missense Mutation | CGC,TGC | R,C 149 | XP_006724572.1 | |
XM_011545564.2 | 3696 | Missense Mutation | CGC,TGC | R,C 178 | XP_011543866.1 | |
XM_017029723.1 | 3696 | Missense Mutation | CGC,TGC | R,C 150 | XP_016885212.1 |