Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCCGACAAGACCTCCCCGGAGC[C/T]CGCCCCCCAGCAGGGCTCCACACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102670 | ||||||||||||||||||||
Literature Links: |
MADCAM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MADCAM1 - mucosal vascular addressin cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130760.2 | 825 | Missense Mutation | CCC,CTC | P,L 272 | NP_570116.2 | |
NM_130762.2 | 825 | Intron | NP_570118.1 |
TPGS1 - tubulin polyglutamylase complex subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |