Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCTTTTGATATTCAGGTGAGAT[A/G]CCAGAGAGCTATAGCTGATTGTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFAP1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AFAP1L1 - actin filament associated protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GRPEL2 - GrpE like 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152407.3 | 365 | Missense Mutation | TAC,TGC | Y,C 80 | NP_689620.2 |
GRPEL2-AS1 - GRPEL2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCYOX1L - prenylcysteine oxidase 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |