Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTACTGCTGGGACTGATTATGTG[C/T]GTGAGCCTGGCGGGTAACGCCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300253 MIM: 300564 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR173 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR173 - G protein-coupled receptor 173 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018969.5 | 377 | Silent Mutation | TGC,TGT | C,C 34 | NP_061842.1 | |
XM_011530798.2 | 377 | Silent Mutation | TGC,TGT | C,C 34 | XP_011529100.1 |
TSPYL2 - TSPY like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |