Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGACGGCGGGGTCCTGGGGCTGAG[C/T]GTGGAGCAGGTGGGCACGCTGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611893 MIM: 190700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4530 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4530 - microRNA 4530 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLEKHG2 - pleckstrin homology and RhoGEF domain containing G2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022835.2 | 1279 | Silent Mutation | AGC,AGT | S,S 141 | NP_073746.2 | |
XM_005259163.2 | 1279 | Silent Mutation | AGC,AGT | S,S 141 | XP_005259220.1 | |
XM_006723334.2 | 1279 | Silent Mutation | AGC,AGT | S,S 82 | XP_006723397.1 | |
XM_011527232.2 | 1279 | Silent Mutation | AGC,AGT | S,S 142 | XP_011525534.1 | |
XM_017027150.1 | 1279 | Silent Mutation | AGC,AGT | S,S 142 | XP_016882639.1 | |
XM_017027151.1 | 1279 | Silent Mutation | AGC,AGT | S,S 142 | XP_016882640.1 |
ZFP36 - ZFP36 ring finger protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |