Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTAATTTTTATTACAGTAAGGT[C/T]GGAAATAACCAGATGTCGTGAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC172 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC172 - coiled-coil domain containing 172 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198515.2 | 665 | Missense Mutation | TCG,TTG | S,L 29 | NP_940917.1 | |
XM_006717820.3 | 665 | Missense Mutation | TCG,TTG | S,L 29 | XP_006717883.1 | |
XM_011539771.2 | 665 | Missense Mutation | TCG,TTG | S,L 29 | XP_011538073.1 | |
XM_017016206.1 | 665 | Missense Mutation | TCG,TTG | S,L 29 | XP_016871695.1 |