Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCAAGACCAGTGCCACGTAGGA[G/T]TCTGCTTCGTGTTTAGCTGCGACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186977 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCTE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCTE3 - t-complex-associated-testis-expressed 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006715554.3 | 675 | Missense Mutation | GAA,GAC | E,D 124 | XP_006715617.1 | |
XM_011536093.2 | 675 | Missense Mutation | GAA,GAC | E,D 184 | XP_011534395.1 |