Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACTGACATGCTGCATTTTTTCAT[C/G]TCTTTCACGCAGCTTGTCTGTTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611176 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC175 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC175 - coiled-coil domain containing 175 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164399.1 | 2485 | Missense Mutation | CAT,GAT | H,D 738 | NP_001157871.1 | |
XM_011537120.2 | 2485 | Missense Mutation | CAT,GAT | H,D 739 | XP_011535422.1 | |
XM_011537122.2 | 2485 | Missense Mutation | CAT,GAT | H,D 665 | XP_011535424.1 | |
XM_011537124.2 | 2485 | Missense Mutation | CAT,GAT | H,D 614 | XP_011535426.1 | |
XM_011537125.2 | 2485 | Intron | XP_011535427.1 | |||
XM_011537130.2 | 2485 | Intron | XP_011535432.1 | |||
XM_017021621.1 | 2485 | Intron | XP_016877110.1 |
JKAMP - JNK1/MAPK8-associated membrane protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |